Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173185.2 | NP_775277.2 | 3 - 4 | 597 | 102 | |
GenBank mRNA | AK090262.1 | - | 3 - 4 | 598 | 102 | |
AK149310.1 | - | 4 - 5 | 615 | 102 | ||
BC064645.1 | - | 2 - 3 | 613 | 5363779 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |