Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029796.1 | - | 3 - 4 | 1561 | 73 | |
BC079845.1 | - | 3 - 4 | 359 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |