Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164147.1 | NP_001157619.1 | 11 - 12 | 1344 | 102 | |
NM_001164148.1 | NP_001157620.1 | 11 - 12 | 1344 | 102 | ||
NM_001164149.1 | NP_001157621.1 | 11 - 12 | 1341 | 102 | ||
NM_001164150.1 | NP_001157622.1 | 11 - 12 | 1344 | 102 | ||
NM_001164151.1 | NP_001157623.1 | 11 - 12 | 1341 | 102 | ||
NM_001164152.1 | NP_001157624.1 | 11 - 12 | 1344 | 102 | ||
NM_001164153.1 | NP_001157625.1 | 11 - 12 | 1341 | 102 | ||
NM_011548.4 | NP_035678.3 | 11 - 12 | 1341 | 102 | ||
GenBank mRNA | AF352579.1 | - | 11 - 12 | 1036 | 102 | |
AK034777.1 | - | 11 - 12 | 1324 | 102 | ||
AK038738.1 | - | 11 - 12 | 1328 | 102 | ||
AK146786.1 | - | 11 - 12 | 1335 | 102 | ||
AK147735.1 | - | 10 - 11 | 1000 | 102 | ||
AK156265.1 | - | 11 - 12 | 1324 | 102 | ||
AK161009.1 | - | 11 - 12 | 1330 | 102 | ||
AK170674.1 | - | 11 - 12 | 102 | |||
BC006860.1 | - | 11 - 12 | 1295 | 2631291 | 102 | |
BC018260.1 | - | 10 - 11 | 969 | 4187386 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |