Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025910.3 | NP_080186.3 | 2 - 3 | 633 | 84 | |
GenBank mRNA | AB177385.1 | - | 1 - 2 | 434 | 84 | |
AK007810.1 | - | 2 - 3 | 603 | 84 | ||
AK013451.1 | - | 2 - 3 | 593 | 84 | ||
AK031671.1 | - | 2 - 3 | 567 | 84 | ||
BC023462.1 | - | 2 - 3 | 888 | 5041558 | 84 | |
BC025109.1 | - | 2 - 3 | 555 | 4236994 | 84 | |
BC058242.1 | - | 2 - 3 | 542 | 6308890 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |