Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028472.2 | NP_082748.1 | 9 - 10 | 1489 | 60 | |
GenBank mRNA | AF454954.1 | - | 9 - 10 | 1489 | 60 | |
AK014221.1 | - | 7 - 8 | 694 | 60 | ||
AY263358.1 | - | 9 - 10 | 1010 | 60 | ||
BC066153.1 | - | 9 - 10 | 994 | 30098453 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |