Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008026.5 | NP_032052.1 | 2 - 3 | 476 | 77 | |
GenBank mRNA | AK042111.1 | - | 2 - 3 | 389 | 77 | |
AK154714.1 | - | 2 - 3 | 393 | 77 | ||
BC138791.1 | - | 2 - 3 | 303 | 8861813 | 77 | |
BC138792.1 | - | 2 - 3 | 303 | 8861814 | 77 | |
X59421.1 | - | 2 - 3 | 476 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |