Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qtrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021888.2 | NP_068688.2 | 7 - 8 | 878 | 73 | |
GenBank mRNA | AB034632.1 | - | 7 - 8 | 812 | 73 | |
AK011586.1 | - | 7 - 8 | 871 | 73 | ||
BC044811.1 | - | 7 - 8 | 872 | 5031330 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |