Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042707.2 | NP_001036172.1 | 3 - 4 | 557 | 66 | |
NM_001042708.2 | NP_001036173.1 | 4 - 5 | 596 | 66 | ||
NM_001042709.2 | NP_001036174.1 | 3 - 4 | 557 | 66 | ||
NM_001277321.1 | NP_001264250.1 | 4 - 5 | 596 | 66 | ||
NM_001277322.1 | NP_001264251.1 | 5 - 6 | 524 | 66 | ||
NM_010561.3 | NP_034691.2 | 4 - 5 | 596 | 66 | ||
GenBank mRNA | AF497751.2 | - | 4 - 5 | 464 | 66 | |
AF497752.2 | - | 3 - 4 | 425 | 66 | ||
AK048096.1 | - | 3 - 4 | 487 | 66 | ||
AK077560.1 | - | 4 - 5 | 522 | 66 | ||
AK128936.1 | - | 4 - 5 | 529 | 66 | ||
AK150276.1 | - | 4 - 5 | 545 | 66 | ||
BC047272.1 | - | 4 - 5 | 516 | 6440312 | 66 | |
BC060599.1 | - | 2 - 3 | 260 | 66 | ||
DQ104405.1 | - | 4 - 5 | 464 | 66 | ||
DQ104406.1 | - | 3 - 4 | 425 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |