Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042707.2 | NP_001036172.1 | 6 - 7 | 974 | 127 | |
NM_001042708.2 | NP_001036173.1 | 7 - 8 | 1013 | 127 | ||
NM_001042709.2 | NP_001036174.1 | 6 - 7 | 974 | 127 | ||
NM_001277321.1 | NP_001264250.1 | 7 - 8 | 1013 | 127 | ||
NM_001277322.1 | NP_001264251.1 | 8 - 9 | 941 | 127 | ||
NM_010561.3 | NP_034691.2 | 7 - 8 | 1013 | 127 | ||
GenBank mRNA | AB221609.1 | - | 5 - 6 | 689 | 127 | |
AF098967.1 | - | 8 - 9 | 890 | 127 | ||
AF497751.2 | - | 7 - 8 | 881 | 127 | ||
AF497752.2 | - | 6 - 7 | 842 | 127 | ||
AK048096.1 | - | 6 - 7 | 904 | 127 | ||
AK077560.1 | - | 7 - 8 | 939 | 127 | ||
AK128936.1 | - | 7 - 8 | 946 | 127 | ||
AK150276.1 | - | 7 - 8 | 962 | 127 | ||
BC047272.1 | - | 7 - 8 | 933 | 6440312 | 127 | |
BC060599.1 | - | 5 - 6 | 677 | 127 | ||
DQ104405.1 | - | 7 - 8 | 881 | 127 | ||
DQ104406.1 | - | 6 - 7 | 842 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |