Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009314.4 | NP_033340.3 | 3 - 4 | 1115 | 67 | |
GenBank mRNA | AK033443.1 | - | 3 - 4 | 798 | 67 | |
BC106959.1 | - | 3 - 4 | 764 | 40039032 | 67 | |
BC106960.2 | - | 2 - 3 | 67 | |||
X62933.1 | - | 3 - 4 | 1115 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |