Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144933.1 | NP_659182.1 | 7 - 8 | 1202 | 68 | |
GenBank mRNA | AK033151.1 | - | 7 - 8 | 1208 | 68 | |
AK038451.1 | - | 7 - 8 | 1242 | 68 | ||
AK049112.1 | - | 7 - 8 | 1242 | 68 | ||
AK142033.1 | - | 7 - 8 | 1241 | 68 | ||
AK151377.1 | - | 7 - 8 | 1242 | 68 | ||
AK178010.1 | - | 1 - 2 | 28 | 68 | ||
BC004062.1 | - | 6 - 7 | 879 | 68 | ||
BC020185.1 | - | 7 - 8 | 1202 | 4505959 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |