Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024139.1 | NP_001019310.1 | 1 - 2 | 1171 | 63 | |
GenBank mRNA | BC094677.1 | - | 1 - 2 | 1171 | 30619053 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |