Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009795.3 | NP_033925.2 | 7 - 8 | 580 | 102 | |
GenBank mRNA | AF058298.1 | - | 6 - 7 | 559 | 102 | |
AK028092.1 | - | 5 - 6 | 326 | 102 | ||
AK132819.1 | - | 6 - 7 | 390 | 102 | ||
BC018352.1 | - | 7 - 8 | 574 | 3156021 | 102 | |
BC090988.1 | - | 7 - 8 | 580 | 30246016 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |