Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011874.2 | NP_036004.2 | 5 - 6 | 659 | 80 | |
GenBank mRNA | AK050677.1 | - | 5 - 6 | 637 | 80 | |
AK077507.1 | - | 5 - 6 | 622 | 80 | ||
AK150989.1 | - | 5 - 6 | 614 | 80 | ||
AK153991.1 | - | 5 - 6 | 642 | 80 | ||
AK160644.1 | - | 5 - 6 | 621 | 80 | ||
AK160718.1 | - | 5 - 6 | 613 | 80 | ||
AK167041.1 | - | 5 - 6 | 604 | 80 | ||
AK167528.1 | - | 5 - 6 | 614 | 80 | ||
AK169224.1 | - | 5 - 6 | 622 | 80 | ||
AK169244.1 | - | 5 - 6 | 1122 | 80 | ||
BC012708.1 | - | 5 - 6 | 620 | 4211677 | 80 | |
BC092265.1 | - | 5 - 6 | 603 | 3671250 | 80 | |
L76223.1 | - | 5 - 6 | 610 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |