Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019459.2 | NP_062332.2 | 16 - 17 | 2339 | 56 | |
GenBank mRNA | AB513652.1 | - | 16 - 17 | 2211 | 56 | |
AF168466.1 | - | 16 - 17 | 2211 | 56 | ||
AF172256.1 | - | 16 - 17 | 2339 | 56 | ||
AF191090.1 | - | 16 - 17 | 2667 | 56 | ||
AK141081.1 | - | 16 - 17 | 2362 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |