Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172142.3 | NP_742154.1 | 9 - 10 | 1234 | 62 | |
GenBank mRNA | AK155969.1 | - | 10 - 11 | 1240 | 62 | |
AK167356.1 | - | 10 - 11 | 1228 | 62 | ||
AY078069.1 | - | 9 - 10 | 1239 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |