Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009561.2 | NP_033587.2 | 3 - 4 | 628 | 77 | |
GenBank mRNA | AK149960.1 | - | 3 - 4 | 423 | 77 | |
BC006618.1 | - | 3 - 4 | 667 | 3967527 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |