Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008408.4 | NP_032434.3 | 8 - 9 | 855 | 74 | |
GenBank mRNA | AK030363.1 | - | 8 - 9 | 919 | 74 | |
AK077877.1 | - | 8 - 9 | 879 | 74 | ||
AK081547.1 | - | 8 - 9 | 911 | 74 | ||
AK129027.1 | - | 8 - 9 | 893 | 74 | ||
AK150640.1 | - | 8 - 9 | 900 | 74 | ||
AK151541.1 | - | 8 - 9 | 911 | 74 | ||
AK151791.1 | - | 8 - 9 | 911 | 74 | ||
AK152242.1 | - | 8 - 9 | 900 | 74 | ||
AK153840.1 | - | 8 - 9 | 856 | 74 | ||
AK153886.1 | - | 8 - 9 | 956 | 74 | ||
AK160037.1 | - | 8 - 9 | 894 | 74 | ||
AK160607.1 | - | 8 - 9 | 856 | 74 | ||
AK168505.1 | - | 8 - 9 | 900 | 74 | ||
AK168937.1 | - | 8 - 9 | 900 | 74 | ||
AK169669.1 | - | 8 - 9 | 879 | 74 | ||
AK217208.1 | - | 1 - 2 | 77 | 74 | ||
AX799090.1 | - | 8 - 9 | 898 | 74 | ||
BC028897.1 | - | 7 - 8 | 806 | 74 | ||
BC037612.1 | - | 8 - 9 | 894 | 5353797 | 74 | |
L34260.1 | - | 8 - 9 | 898 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |