Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ei24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199494.1 | NP_001186423.1 | 2 - 3 | 285 | 75 | |
NM_007915.5 | NP_031941.1 | 2 - 3 | 169 | 75 | ||
GenBank mRNA | AK145018.1 | - | 2 - 3 | 246 | 75 | |
AK147776.1 | - | 2 - 3 | 198 | 75 | ||
AK159927.1 | - | 2 - 3 | 227 | 75 | ||
AK172233.1 | - | 2 - 3 | 249 | 75 | ||
AK206494.1 | - | 2 - 3 | 124 | 75 | ||
BU148129.1 | - | 2 - 3 | 238 | 75 | ||
BY282890.1 | - | 2 - 3 | 286 | 75 | ||
U41751.2 | - | 2 - 3 | 173 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mus musculus Probe spans exons |