Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139001.2 | NP_620570.2 | 6 - 7 | 4746 | 57 | |
GenBank mRNA | AK140986.1 | - | 1 - 2 | 154 | 57 | |
AK220559.1 | - | 4 - 5 | 1667 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |