Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025837.2 | NP_080113.1 | 3 - 4 | 381 | 64 | |
GenBank mRNA | AF244360.1 | - | 3 - 4 | 363 | 64 | |
AK003289.1 | - | 1 - 2 | 141 | 64 | ||
AK003896.1 | - | 3 - 4 | 381 | 64 | ||
AK132777.1 | - | 3 - 4 | 382 | 64 | ||
BC009068.1 | - | 3 - 4 | 359 | 2650791 | 64 | |
BC051163.1 | - | 3 - 4 | 359 | 6466145 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |