Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199222.3 | NP_954692.2 | 11 - 12 | 1361 | 84 | |
GenBank mRNA | AB211986.1 | - | 11 - 12 | 1282 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |