Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dixdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178118.2 | NP_835219.1 | 1 - 2 | 204 | 71 | |
GenBank mRNA | AK016205.1 | - | 1 - 2 | 159 | 71 | |
AK164424.1 | - | 1 - 2 | 205 | 71 | ||
AY549884.1 | - | 1 - 2 | 65 | 71 | ||
BC048182.1 | - | 1 - 2 | 195 | 6826299 | 71 | |
BC063085.1 | - | 1 - 2 | 195 | 6826299 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |