Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199195.1 | NP_954665.1 | 4 - 5 | 409 | 75 | |
GenBank mRNA | AK089921.1 | - | 4 - 5 | 487 | 75 | |
BC064099.1 | - | 4 - 5 | 409 | 1246285 | 75 | |
L16992.1 | - | 4 - 5 | 481 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |