Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172896.2 | NP_766484.1 | 1 - 2 | 145 | 80 | |
GenBank mRNA | AK087843.1 | - | 1 - 2 | 146 | 80 | |
AY355341.1 | - | 1 - 2 | 146 | 80 | ||
BC141342.1 | - | 1 - 2 | 45 | 9055954 | 80 | |
BC141343.1 | - | 1 - 2 | 45 | 9055955 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |