Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182785.3 | NP_877586.1 | 1 - 2 | 162 | 86 | |
GenBank mRNA | AK077071.1 | - | 1 - 2 | 163 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |