Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039507.2 | NP_001034596.1 | 7 - 8 | 1524 | 77 | |
NM_010719.5 | NP_034849.2 | 7 - 8 | 1960 | 77 | ||
GenBank mRNA | AK019846.1 | - | 7 - 8 | 2361 | 77 | |
AK029984.1 | - | 7 - 8 | 1963 | 77 | ||
AK169858.1 | - | 7 - 8 | 1519 | 77 | ||
BC021642.1 | - | 7 - 8 | 1520 | 4188567 | 77 | |
BC053502.1 | - | 7 - 8 | 1584 | 77 | ||
U08188.1 | - | 6 - 7 | 2045 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |