Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104617.1 | NP_001098087.1 | 1 - 2 | 488 | 108 | |
NM_009041.3 | NP_033067.2 | 1 - 2 | 488 | 108 | ||
GenBank mRNA | AK016711.1 | - | 1 - 2 | 159 | 108 | |
AK088292.1 | - | 1 - 2 | 195 | 108 | ||
AK146423.1 | - | 1 - 2 | 146 | 108 | ||
AK156370.1 | - | 1 - 2 | 174 | 108 | ||
AK168449.1 | - | 1 - 2 | 159 | 108 | ||
BC002041.1 | - | 1 - 2 | 133 | 108 | ||
BC026786.1 | - | 1 - 2 | 161 | 108 | ||
BC034438.1 | - | 1 - 2 | 314 | 108 | ||
BC053417.1 | - | 1 - 2 | 162 | 6707873 | 108 | |
X60672.1 | - | 1 - 2 | 101 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |