Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281954.1 | NP_001268883.1 | 4 - 5 | 523 | 132 | |
NM_019465.4 | NP_062338.3 | 4 - 5 | 523 | 132 | ||
GenBank mRNA | AK005719.1 | - | 2 - 3 | 246 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |