Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081688.2 | NP_001075157.2 | 6 - 7 | 680 | 84 | |
GenBank mRNA | AB109391.1 | - | 6 - 7 | 791 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |