Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prtg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175485.4 | NP_780694.3 | 5 - 6 | 974 | 75 | |
GenBank mRNA | AK083540.1 | - | 3 - 4 | 410 | 75 | |
AY630257.1 | - | 5 - 6 | 1044 | 75 | ||
BC116336.1 | - | 3 - 4 | 407 | 75 | ||
BC116337.1 | - | 3 - 4 | 407 | 75 | ||
DQ360114.1 | - | 5 - 6 | 974 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |