Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripply2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037907.1 | NP_001032996.1 | 2 - 3 | 172 | 86 | |
GenBank mRNA | AB212223.1 | - | 2 - 3 | 172 | 86 | |
AK081203.1 | - | 1 - 2 | 314 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |