Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013932.4 | NP_038960.2 | 4 - 5 | 379 | 87 | |
GenBank mRNA | AF142630.3 | - | 4 - 5 | 310 | 87 | |
AK050693.1 | - | 4 - 5 | 374 | 87 | ||
AK078340.1 | - | 4 - 5 | 338 | 87 | ||
BC024852.1 | - | 4 - 5 | 323 | 5358404 | 87 | |
BC061130.1 | - | 4 - 5 | 334 | 6774579 | 87 | |
CF104207.1 | - | 4 - 5 | 379 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |