Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028040.2 | NP_082316.1 | 4 - 5 | 662 | 89 | |
GenBank mRNA | AK010322.1 | - | 4 - 5 | 679 | 89 | |
AK156574.1 | - | 4 - 5 | 679 | 89 | ||
BC061256.1 | - | 4 - 5 | 663 | 30251586 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |