Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009178.3 | NP_033204.2 | 3 - 4 | 277 | 76 | |
GenBank mRNA | AB061305.1 | - | 3 - 4 | 225 | 76 | |
BC011121.1 | - | 3 - 4 | 222 | 3497996 | 76 | |
BC050773.1 | - | 3 - 4 | 277 | 6587379 | 76 | |
D28941.1 | - | 3 - 4 | 232 | 76 | ||
X95809.1 | - | 2 - 3 | 104 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |