Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019727.2 | NP_062701.2 | 11 - 12 | 1343 | 85 | |
GenBank mRNA | AB019214.1 | - | 11 - 12 | 1283 | 85 | |
AF154120.1 | - | 11 - 12 | 1347 | 85 | ||
AK076198.1 | - | 3 - 4 | 84 | |||
AK154079.1 | - | 11 - 12 | 1256 | 85 | ||
AK155746.1 | - | 11 - 12 | 1231 | 85 | ||
AK161241.1 | - | 11 - 12 | 1324 | 85 | ||
AK167177.1 | - | 11 - 12 | 1298 | 85 | ||
AK167993.1 | - | 11 - 12 | 1238 | 85 | ||
AK168742.1 | - | 11 - 12 | 1336 | 85 | ||
AK192200.1 | - | 3 - 4 | 345 | 85 | ||
BC066044.1 | - | 11 - 12 | 1300 | 6807363 | 85 | |
BC066189.1 | - | 11 - 12 | 1234 | 30471747 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |