Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019727.2 | NP_062701.2 | 6 - 7 | 769 | 88 | |
GenBank mRNA | AB019214.1 | - | 6 - 7 | 709 | 88 | |
AF154120.1 | - | 6 - 7 | 773 | 88 | ||
AK149850.1 | - | 6 - 7 | 681 | 88 | ||
AK154079.1 | - | 6 - 7 | 682 | 88 | ||
AK155746.1 | - | 6 - 7 | 657 | 88 | ||
AK161241.1 | - | 6 - 7 | 750 | 88 | ||
AK167177.1 | - | 6 - 7 | 724 | 88 | ||
AK167993.1 | - | 6 - 7 | 664 | 88 | ||
AK168742.1 | - | 6 - 7 | 762 | 88 | ||
AK219007.1 | - | 4 - 5 | 299 | 88 | ||
BC066044.1 | - | 6 - 7 | 726 | 6807363 | 88 | |
BC066189.1 | - | 6 - 7 | 660 | 30471747 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |