Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144922.2 | NP_659171.1 | 3 - 4 | 658 | 99 | |
NM_178089.2 | NP_835190.1 | 3 - 4 | 639 | 99 | ||
GenBank mRNA | AK042090.1 | - | 3 - 4 | 639 | 99 | |
AK087545.1 | - | 3 - 4 | 628 | 99 | ||
AK143666.1 | - | 3 - 4 | 701 | 99 | ||
AK155604.1 | - | 3 - 4 | 669 | 99 | ||
BC021506.1 | - | 3 - 4 | 682 | 5347099 | 99 | |
BC078642.1 | - | 3 - 4 | 582 | 6329630 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |