Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172289.4 | NP_758493.2 | 1 - 2 | 69 | 92 | |
GenBank mRNA | AK032074.1 | - | 1 - 2 | 70 | 92 | |
AK052277.1 | - | 1 - 2 | 70 | 92 | ||
AK150167.1 | - | 1 - 2 | 101 | 92 | ||
BC115964.1 | - | 1 - 2 | 70 | 40057255 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |