Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011139.2 | NP_035269.2 | 2 - 3 | 169 | 80 | |
GenBank mRNA | AK153697.1 | - | 259 | 80 | ||
L14677.1 | - | 2 - 3 | 167 | 80 | ||
Z18537.1 | - | 2 - 3 | 170 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |