Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013487.3 | NP_038515.3 | 2 - 3 | 380 | 84 | |
GenBank mRNA | AK042172.1 | - | 2 - 3 | 382 | 84 | |
AK050629.1 | - | 2 - 3 | 369 | 84 | ||
AK153843.1 | - | 2 - 3 | 345 | 84 | ||
AK154064.1 | - | 2 - 3 | 342 | 84 | ||
BC062807.1 | - | 2 - 3 | 285 | 1366111 | 84 | |
EF382667.1 | - | 2 - 3 | 280 | 84 | ||
HM536173.1 | - | 2 - 3 | 280 | 84 | ||
X02339.1 | - | 2 - 3 | 248 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |