Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027865.2 | NP_082141.1 | 2 - 3 | 129 | 67 | |
GenBank mRNA | AK002841.1 | - | 2 - 3 | 130 | 67 | |
AK090375.1 | - | 1 - 2 | 535 | 67 | ||
AK165199.1 | - | 2 - 3 | 161 | 67 | ||
AK165827.1 | - | 2 - 3 | 114 | 67 | ||
BC060057.1 | - | 2 - 3 | 149 | 5696102 | 67 | |
BC060243.1 | - | 2 - 3 | 149 | 5696102 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |