Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145403.2 | NP_663378.1 | 1 - 2 | 292 | 100 | |
GenBank mRNA | AK078890.1 | - | 1 - 2 | 307 | 100 | |
AY043240.1 | - | 1 - 2 | 292 | 100 | ||
BC021368.1 | - | 1 - 2 | 241 | 5030266 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |