Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198962.3 | NP_945200.1 | 5 - 6 | 1055 | 90 | |
GenBank mRNA | AK038551.1 | - | 5 - 6 | 1056 | 90 | |
AK048781.1 | - | 5 - 6 | 1055 | 90 | ||
AK079572.1 | - | 5 - 6 | 1088 | 90 | ||
AY336084.1 | - | 5 - 6 | 1023 | 90 | ||
AY336085.1 | - | 5 - 6 | 1023 | 90 | ||
BC140957.1 | - | 5 - 6 | 1051 | 9055569 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |