Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019689.2 | NP_062663.1 | 2 - 3 | 699 | 134 | |
GenBank mRNA | AF116847.1 | - | 2 - 3 | 699 | 134 | |
AK148073.1 | - | 3 - 4 | 796 | 134 | ||
BC049776.1 | - | 3 - 4 | 921 | 134 | ||
BC150755.1 | - | 2 - 3 | 693 | 9087666 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |