Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019689.2 | NP_062663.1 | 5 - 6 | 1270 | 82 | |
GenBank mRNA | AF116847.1 | - | 5 - 6 | 1270 | 82 | |
AK139455.1 | - | 7 - 8 | 1452 | 82 | ||
AK148073.1 | - | 6 - 7 | 1368 | 82 | ||
BC145330.1 | - | 5 - 6 | 1334 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |