Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028854.3 | NP_083130.1 | 5 - 6 | 519 | 74 | |
GenBank mRNA | AK007086.1 | - | 5 - 6 | 607 | 74 | |
BC100468.1 | - | 5 - 6 | 585 | 30916687 | 74 | |
BC132329.1 | - | 5 - 6 | 519 | 40130606 | 74 | |
BC138332.1 | - | 5 - 6 | 519 | 8861352 | 74 | |
BC145239.1 | - | 5 - 6 | 519 | 9053791 | 74 | |
BC145240.1 | - | 5 - 6 | 519 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |