Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000003280.mm.1 |
NM_001039051.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003280.mm.1 |
NM_001039051.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003280.mm.1 |
NM_008863.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003280.mm.1 |
NM_008863.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003280.mm.1 |
TC1000000506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001039050.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001039050.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001039051.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001039051.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001039052.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001039052.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001039053.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001039053.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001190402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_001190402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_008863.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
NM_008863.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003291.mm.1 |
TC1000000506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003281.mm.1 |
NM_001039051.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003281.mm.1 |
NM_001039051.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003281.mm.1 |
NM_008863.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003281.mm.1 |
NM_008863.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003281.mm.1 |
TC1000000506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003282.mm.1 |
NM_001039051.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003282.mm.1 |
NM_001039051.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003282.mm.1 |
NM_008863.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003282.mm.1 |
NM_008863.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003282.mm.1 |
TC1000000506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003278.mm.1 |
NM_001039051.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003278.mm.1 |
NM_001039051.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003278.mm.1 |
NM_008863.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003278.mm.1 |
NM_008863.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003278.mm.1 |
TC1000000506.mm.1 |
MTA TC |