Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008297.3 | NP_032323.3 | 8 - 9 | 958 | 134 | |
GenBank mRNA | AK052270.1 | - | 8 - 9 | 134 | ||
AK053976.1 | - | 8 - 9 | 991 | 134 | ||
BC018414.1 | - | 8 - 9 | 880 | 3990765 | 134 | |
X61754.1 | - | 8 - 9 | 992 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |