Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013718.2 | NP_038746.1 | 1 - 2 | 133 | 56 | |
GenBank mRNA | AF041433.1 | - | 1 - 2 | 124 | 56 | |
AK151747.1 | - | 1 - 2 | 145 | 56 | ||
AK168518.1 | - | 1 - 2 | 173 | 56 | ||
BC003736.1 | - | 1 - 2 | 133 | 3490979 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |